missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Pyrrole-2-carboxylate 98.0+%, TCI America™

Catalog No. E10305G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E10305G 5 g
E103025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E10305G Supplier TCI America Supplier No. E10305G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2199-43-1
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00817049
InChI Key PAEYAKGINDQUCT-UHFFFAOYSA-N
Synonym ethyl pyrrole-2-carboxylate, ethyl 2-pyrrolecarboxylate, ethyl pyrrole-2-carbonate, ethylpyrrole-2-carboxylate, 1h-pyrrole-2-carboxylic acid, ethyl ester, 2-ethoxycarbonyl-1h-pyrrole, pyrrole-2-carboxylic acid, ethyl ester, 1h-pyrrole-2-carboxylic acid ethyl ester, pyrrole-2-carboxylic acid ethyl ester, ethyl1h-pyrrole-2-carboxylate
PubChem CID 255670
IUPAC Name ethyl 1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=CC=CN1

Specifications

Melting Point 41°C
Boiling Point 235°C
Quantity 5 g
Formula Weight 139.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Pyrrole-2-carboxylate