accessibility menu, dialog, popup

UserName

CAS RN 2198-66-5

CH3H3CCH3OHBr

CAS RN 2198-66-5

IUPAC Name: 2-bromo-4-tert-butylphenol
Synonyms: 2-bromo-4-tert-butylphenol
Molecular Weight (g/mol): 229.12
Molecular Formula: C10H13BrO
InChi Key: FFRLMQPMGIMHHQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(O)C(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results