accessibility menu, dialog, popup

UserName

CAS RN 21963-41-7

IUPAC Name: cyclohexane-1,2-dicarboxylic acid
Synonyms: cyclohexane-1,2-dicarboxylic acid
Molecular Weight (g/mol): 172.18
Molecular Formula: C8H12O4
InChi Key: QSAWQNUELGIYBC-UHFFFAOYNA-N
SMILES: OC(=O)C1CCCCC1C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)