missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-Cyclohexane-1,2-dicarboxylic acid, 98+%

Catalog No. AAH5229006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5229006 5 g
AAH5229003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5229006 Supplier Thermo Scientific Chemicals Supplier No. H5229006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21963-41-7 | C8H12O4 | 172.18 g/mol

A series of new C2-symmetric (1S,2S)-cyclohexane-1,2-dicarboxamides was synthesized from (1S,2S)-Cyclohexane-1,2-dicarboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A series of new C2-symmetric (1S,2S)-cyclohexane-1,2-dicarboxamides was synthesized from (1S,2S)-Cyclohexane-1,2-dicarboxylic acid.

Solubility
Soluble in acetone(almost transparency).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 21963-41-7
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00151258,MFCD00001464
InChI Key QSAWQNUELGIYBC-UHFFFAOYNA-N
Synonym 1s,2s-cyclohexane-1,2-dicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, trans, 1,2-cyclohexanedicarboxylic acid, 1s,2s, 1,2-cyclohexanedicarboxylicacid, 1s,2s, pubchem19269, trans-hexahydrophthalic acid, 1alpha,2beta-cyclohexanedicarboxylic acid, 1s,2s-1,2-cyclohexanedicarboxylicacid
PubChem CID 75315
IUPAC Name (1S,2S)-cyclohexane-1,2-dicarboxylic acid
SMILES OC(=O)C1CCCCC1C(O)=O

Specifications

Quantity 5 g
Solubility Information Soluble in acetone(almost transparency).
Formula Weight 172.18
Percent Purity ≥98%
Chemical Name or Material (1S,2S)-Cyclohexane-1,2-dicarboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only