accessibility menu, dialog, popup

UserName

CAS RN 2179-57-9

H2CSSCH2

CAS RN 2179-57-9

IUPAC Name: 3-(prop-2-en-1-yldisulfanyl)prop-1-ene
Synonyms: garlicin allitin
Molecular Weight (g/mol): 146.27
Molecular Formula: C6H10S2
InChi Key: PFRGXCVKLLPLIP-UHFFFAOYSA-N
SMILES: C=CCSSCC=C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)