CAS RN 21743-75-9
CAS RN 21743-75-9
IUPAC Name:
2-(2H-1,2,3,4-tetrazol-5-yl)acetic acid
Synonyms:
2H-1,2,3,4-tetrazol-5-ylacetic acid
Molecular Weight (g/mol):
128.09
Molecular Formula:
C3H4N4O2
InChi Key:
JUNAPQMUUHSYOV-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=NNN=N1
1
–
1
of
1
results
1
1H-Tetrazole-5-acetic Acid 98.0+%, TCI America™
CAS: 21743-75-9 Molecular Formula: C3H4N4O2 Molecular Weight (g/mol): 128.091 MDL Number: MFCD03094010 InChI Key: JUNAPQMUUHSYOV-UHFFFAOYSA-N PubChem CID: 89031 IUPAC Name: 2-(2H-tetrazol-5-yl)acetic acid SMILES: C(C1=NNN=N1)C(=O)O