missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Tetrazole-5-acetic Acid 98.0+%, TCI America™
$367.22 - $1,780.17
Chemical Identifiers
| CAS | 21743-75-9 |
|---|---|
| Molecular Formula | C3H4N4O2 |
| Molecular Weight (g/mol) | 128.091 |
| MDL Number | MFCD03094010 |
| InChI Key | JUNAPQMUUHSYOV-UHFFFAOYSA-N |
| PubChem CID | 89031 |
| IUPAC Name | 2-(2H-tetrazol-5-yl)acetic acid |
| SMILES | C(C1=NNN=N1)C(=O)O |
Chemical Identifiers
| 21743-75-9 | |
| 128.091 | |
| JUNAPQMUUHSYOV-UHFFFAOYSA-N | |
| 2-(2H-tetrazol-5-yl)acetic acid |
| C3H4N4O2 | |
| MFCD03094010 | |
| 89031 | |
| C(C1=NNN=N1)C(=O)O |
Specifications
| 21743-75-9 | |
| White | |
| MFCD03094010 | |
| 1325 | |
| C(C1=NNN=N1)C(=O)O | |
| 128.091 | |
| 128.09 | |
| Crystalline Powder |
| 177°C | |
| C3H4N4O2 | |
| 5 g | |
| JUNAPQMUUHSYOV-UHFFFAOYSA-N | |
| 2-(2H-tetrazol-5-yl)acetic acid | |
| 89031 | |
| ≥98.0% (GC,T) | |
| 1H-Tetrazole-5-acetic Acid |
Safety and Handling
TSCA : No