accessibility menu, dialog, popup

UserName

CAS RN 21278-86-4

IUPAC Name: 2-(pyridin-4-yl)-1,3-thiazole-4-carboxylate
Synonyms: 2-(pyridin-4-yl)-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol): 205.21
Molecular Formula: C9H5N2O2S
InChi Key: COOQMBOJAAZEIR-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CSC(=N1)C1=CC=NC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results