missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Pyridyl)thiazole-4-carboxylic acid, 97%

Catalog No. AAB2045706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2045706 5 g
AAB2045703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2045706 Supplier Thermo Scientific Chemicals Supplier No. B2045706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 21278-86-4 | C9H5N2O2S | 205.21 g/mol

2-(4-Pyridyl)thiazole-4-carboxylic acid, is used as pharmaceutical intermediate and also used in the field of organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Pyridyl)thiazole-4-carboxylic acid, is used as pharmaceutical intermediate and also used in the field of organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents, strong acids, strong bases.

Chemical Identifiers

CAS 21278-86-4
Molecular Formula C9H5N2O2S
Molecular Weight (g/mol) 205.21
MDL Number MFCD00171745
InChI Key COOQMBOJAAZEIR-UHFFFAOYSA-M
Synonym 2-pyridin-4-yl thiazole-4-carboxylic acid, 2-4-pyridyl thiazole-4-carboxylic acid, 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid, 2-4-pyridyl-1,3-thiazole-4-carboxylic acid, 2-4-pyridinyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-4-yl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-4-pyridinyl, acmc-1cr1v, 4-thiazolecarboxylic acid, 2-4-pyridinyl, 4-carboxy-2-4-pyridyl thiazole
PubChem CID 716091
SMILES [O-]C(=O)C1=CSC(=N1)C1=CC=NC=C1

Specifications

Melting Point >300°C (decomposition)
Quantity 5 g
Solubility Information Insoluble in water.
IUPAC Name 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid
Formula Weight 206.22
Percent Purity 97%
Chemical Name or Material 2-(4-Pyridyl)thiazole-4-carboxylic acid
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only