accessibility menu, dialog, popup

UserName

CAS RN 2127-09-5

IUPAC Name: 5-nitro-1,2-dihydropyridine-2-thione
Synonyms: 5-nitro-1H-pyridine-2-thione
Molecular Weight (g/mol): 156.16
Molecular Formula: C5H4N2O2S
InChi Key: BHQUBONFIYNJDA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CNC(=S)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results