accessibility menu, dialog, popup

UserName

CAS RN 21087-64-9

H3CSNNONH2NCH3H3CCH3

CAS RN 21087-64-9

IUPAC Name: 4-amino-6-tert-butyl-3-(methylsulfanyl)-4,5-dihydro-1,2,4-triazin-5-one
Synonyms: sencor sencorex L.F. metribuzin bayer 6159H sencorex senkor metribuzine epa pesticide chemical code 101101 bayer 6443H sencor 4F
Molecular Weight (g/mol): 214.29
Molecular Formula: C8H14N4OS
InChi Key: FOXFZRUHNHCZPX-UHFFFAOYSA-N
SMILES: CSC1=NN=C(C(=O)N1N)C(C)(C)C
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)