missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ Metribuzin, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022772
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022772 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022772 Supplier Merck Emd Millipore Supplier No. 7529650MG

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

Specifications

CAS 21087-64-9
Molecular Formula C8H14N4OS
Linear Formula C8H14N4OS
MDL Number MFCD00055525
UN Number UN 3077 9 / PGIII
Quantity 50 mg
Formula Weight 214.29
Grade TraceCERT™ Analytical Standard

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC