accessibility menu, dialog, popup

UserName

NH2ClF

CAS RN 2106-05-0

IUPAC Name: 5-chloro-2-fluoroaniline
Synonyms: 5-chloro-2-fluoroaniline
Molecular Weight (g/mol): 145.56
Molecular Formula: C6H5ClFN
InChi Key: JCYROOANFKVAIB-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CC=C1F
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)