missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluoroaniline, 97%

Catalog No. AAB2095118
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2095118 50 g
AAB2095109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2095118 Supplier Thermo Scientific Chemicals Supplier No. B2095118
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2106-05-0 | C6H5ClFN | 145.561 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2106-05-0
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.561
MDL Number MFCD00069416
InChI Key JCYROOANFKVAIB-UHFFFAOYSA-N
Synonym 2-fluoro-5-chloroaniline, 5-chloro-2-fluorophenylamine, benzenamine, 5-chloro-2-fluoro, buttpark 44\01-98, 5-chlor-2-fluoranilin, pubchem1515, 5-chloro-2-fluoroani, acmc-209fh5, 5-chloro-2-fluoro-aniline, 5-chloro-2-fluorobenzenamine
PubChem CID 75015
IUPAC Name 5-chloro-2-fluoroaniline
SMILES C1=CC(=C(C=C1Cl)N)F

Specifications

Density 1.3
Quantity 50 g
Odor Characteristic
Refractive Index 1.56
UN Number UN2810
Boiling Point 102°C to 105°C (20 mmHg)
Formula Weight 145.56
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-fluoroaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 218-284-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only