missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-fluoroaniline 97.0+%, TCI America™

Catalog No. C15965G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15965G 5 g
C159625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C15965G Supplier TCI America Supplier No. C15965G

Chemical Identifiers

CAS 2106-05-0
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.561
MDL Number MFCD00069416
InChI Key JCYROOANFKVAIB-UHFFFAOYSA-N
Synonym 2-fluoro-5-chloroaniline, 5-chloro-2-fluorophenylamine, benzenamine, 5-chloro-2-fluoro, buttpark 44\01-98, 5-chlor-2-fluoranilin, pubchem1515, 5-chloro-2-fluoroani, acmc-209fh5, 5-chloro-2-fluoro-aniline, 5-chloro-2-fluorobenzenamine
PubChem CID 75015
IUPAC Name 5-chloro-2-fluoroaniline
SMILES C1=CC(=C(C=C1Cl)N)F

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 105°C
Formula Weight 145.56
Percent Purity ≥97.0% (GC)
Chemical Name or Material 5-Chloro-2-fluoroaniline