accessibility menu, dialog, popup

UserName

NH2ClF

CAS RN 2106-04-9

IUPAC Name: 3-chloro-2-fluoroaniline
Synonyms: 3-chloro-2-fluoroaniline
Molecular Weight (g/mol): 145.56
Molecular Formula: C6H5ClFN
InChi Key: XWBTZHDDWRNOQH-UHFFFAOYSA-N
SMILES: NC1=CC=CC(Cl)=C1F
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)