missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluoroaniline 98.0+%, TCI America™

Catalog No. C16595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16595G 5 g
C165925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C16595G Supplier TCI America Supplier No. C16595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2106-04-9
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00069415
InChI Key XWBTZHDDWRNOQH-UHFFFAOYSA-N
Synonym 2-fluoro-3-chloroaniline, benzenamine, 3-chloro-2-fluoro, 3-chloro-2-fluorophenylamine, pubchem1529, 3,4-chlorofluoroaniline, 3-chloro-2 fluoroaniline, 2-fluoro-3-chloro-aniline, 3-chloro-2-fluoro aniline, 3-chloro-2-fluoro-aniline, ksc493k4l
PubChem CID 75014
IUPAC Name 3-chloro-2-fluoroaniline
SMILES C1=CC(=C(C(=C1)Cl)F)N

Specifications

Quantity 5 g
Physical Form Clear Liquid at 20°C
Boiling Point 99°C
Formula Weight 145.56
Percent Purity ≥98.0% (GC)
Chemical Name or Material 3-Chloro-2-fluoroaniline