missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluoroaniline, 97%

Catalog No. AAH5114422
Change view
Click to view available options
Quantity:
5 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5114422 100 g
AAH5114406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5114422 Supplier Thermo Scientific Chemicals Supplier No. H5114422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2106-04-9 | C8H11Cl2N5 | 248.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2106-04-9
MDL Number MFCD00069415

Specifications

Density 1.324
Quantity 100 g
Odor Characteristic
Refractive Index 1.564
Molecular Formula C8H11Cl2N5
UN Number UN2810
Synonym 1-3-chlorophenyl biguanide hydrochloride, m-chlorophenylbiguanide hydrochloride, 1-m-chlorophenyl-biguanide hydrochloride, m-cpbg hydrochloride, 1-m-chlorophenyl biguanide hydrochloride, 1-carbamimidamido-n-3-chlorophenyl methanimidamide hydrochloride, imidodicarbonimidic diamide, n-3-chlorophenyl-, monohydrochloride, n-m-chlorophenylbiguanide hydrochloride, biguanide, 1-m-chlorophenyl-, monohydrochloride, n-3-chlorophenyl imidodicarbonimidic diamide hydrochloride
Boiling Point 214°C
InChI Key FOWAIJYHRWFTHR-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)Cl)N=C(N)N=C(N)N.Cl
IUPAC Name 2-(3-chlorophenyl)-1-(diaminomethylidene)guanidine;hydrochloride
Molecular Weight (g/mol) 248.11
PubChem CID 2730228
Formula Weight 145.56
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluoroaniline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only