accessibility menu, dialog, popup

UserName

NH2FCl

CAS RN 2106-02-7

IUPAC Name: 2-chloro-4-fluoroaniline
Synonyms: 2-chloro-4-fluoroaniline
Molecular Weight (g/mol): 145.56
Molecular Formula: C6H5ClFN
InChi Key: XRAKCYJTJGTSMM-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C=C1Cl
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)