missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoroaniline, 97%

Catalog No. AAA1549222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1549222 100 g
AAA1549206 5 g
AAA1549214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1549222 Supplier Thermo Scientific Chemicals Supplier No. A1549222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2106-02-7 | C6H5ClFN | 145.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2106-02-7
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00042530

Specifications

Density 1.219
Quantity 100 g
Odor Characteristic
Refractive Index 1.554
UN Number UN2810
Beilstein 2802559
Synonym 2-fluoro-3-chloroaniline, benzenamine, 3-chloro-2-fluoro, 3-chloro-2-fluorophenylamine, pubchem1529, 3,4-chlorofluoroaniline, 3-chloro-2 fluoroaniline, 2-fluoro-3-chloro-aniline, 3-chloro-2-fluoro aniline, 3-chloro-2-fluoro-aniline, ksc493k4l
Boiling Point 191°C to 192°C
InChI Key XWBTZHDDWRNOQH-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)Cl)F)N
IUPAC Name 3-chloro-2-fluoroaniline
PubChem CID 75014
Formula Weight 145.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluoroaniline
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber 218-282-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only