missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-fluoroaniline 98.0+%, TCI America™

Catalog No. C116125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C116125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C116125G Supplier TCI America Supplier No. C116125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2106-02-7
Molecular Formula C6H5ClFN
Molecular Weight (g/mol) 145.56
MDL Number MFCD00042530
InChI Key XRAKCYJTJGTSMM-UHFFFAOYSA-N
Synonym benzenamine, 2-chloro-4-fluoro, 2-chloro-4-fluorobenzenamine, 2-chloro-4-fluorophenylamine, 2-chloro-4-fluoro-phenylamine, pubchem2943, 2-chloro-4fluoroaniline, 2-chloro-4-fluroaniline, timtec-bb sbb004110, 4-fluoro-2-chloroaniline, acmc-1cj1h
PubChem CID 75013
IUPAC Name 2-chloro-4-fluoroaniline
SMILES C1=CC(=C(C=C1F)Cl)N

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 120°C
Formula Weight 145.56
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Chloro-4-fluoroaniline