accessibility menu, dialog, popup

UserName

CAS RN 2104695-66-9

H3COOCH3BrOSiCH3CH3H3C

CAS RN 2104695-66-9

IUPAC Name: (2-bromo-4,5-dimethoxyphenoxy)trimethylsilane
Synonyms: 2-bromo-4,5-dimethoxyphenoxytrimethylsilane
Molecular Weight (g/mol): 305.24
Molecular Formula: C11H17BrO3Si
InChi Key: SDYUNRUWGJADII-UHFFFAOYSA-N
SMILES: COC1=CC(Br)=C(O[Si](C)(C)C)C=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results