missing translation for 'onlineSavingsMsg'
Learn More

(2-Bromo-4,5-dimethoxyphenoxy)trimethylsilane 96.0+%, TCI America™

Catalog No. B52981G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B52981G Supplier TCI America Supplier No. B52981G
Only null left

Chemical Identifiers

CAS 2104695-66-9
Molecular Formula C11H17BrO3Si
Molecular Weight (g/mol) 305.243
InChI Key SDYUNRUWGJADII-UHFFFAOYSA-N
PubChem CID 132274843
IUPAC Name (2-bromo-4,5-dimethoxyphenoxy)-trimethylsilane
SMILES COC1=CC(=C(C=C1OC)Br)O[Si](C)(C)C

Specifications

Quantity 1 g
Formula Weight 305.24
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (2-Bromo-4,5-dimethoxyphenoxy)trimethylsilane
TSCA No
Recommended Storage Freezer