accessibility menu, dialog, popup

UserName

NFNH2

CAS RN 210169-05-4

IUPAC Name: 5-fluoropyridin-3-amine
Synonyms: 5-fluoropyridin-3-amine
Molecular Weight (g/mol): 112.11
Molecular Formula: C5H5FN2
InChi Key: ZRORIJXOWXYPMO-UHFFFAOYSA-N
SMILES: NC1=CC(F)=CN=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)