missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-fluoropyridine, 98%, Thermo Scientific Chemicals

Catalog No. AAH3411106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3411106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3411106 Supplier Thermo Scientific Chemicals Supplier No. H3411106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 210169-05-4
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.107
MDL Number MFCD00234073
InChI Key ZRORIJXOWXYPMO-UHFFFAOYSA-N
Synonym 3-amino-5-fluoropyridine, 5-amino-3-fluoropyridine, 3-pyridinamine, 5-fluoro, 5-fluoro-3-pyridylamine, 5-fluoro-3-pyridinamine, 3-fluoro-5-aminopyridine, 5-fluoropyridin-3-ylamine, 3-amine-5-fluoro-pyridin, 3-amino-5-fluoro-pyridin, 3-amine-5-fluoro-pyridine
PubChem CID 574450
IUPAC Name 5-fluoropyridin-3-amine
SMILES C1=C(C=NC=C1F)N

Specifications

Melting Point 85°C to 89°C
Quantity 5 g
Formula Weight 112.11
Percent Purity 98%
Chemical Name or Material 3-Amino-5-fluoropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only