accessibility menu, dialog, popup

UserName

CH3NBr

CAS RN 20965-20-2

IUPAC Name: 3-bromobutanenitrile
Synonyms: 3-bromobutanenitrile
Molecular Weight (g/mol): 148
Molecular Formula: C4H6BrN
InChi Key: MBXSHBIQMDKTEW-UHFFFAOYNA-N
SMILES: CC(Br)CC#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results