missing translation for 'onlineSavingsMsg'
Learn More

Digoxin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018516
Change view
Click to view available options
Quantity:
300 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018516 300 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018516 Supplier Merck Emd Millipore Supplier No. PHR1771300MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Digoxin is one of the digitalis glycosides, extensively utilized for the treatment of congestive heart failure (CHF). It exhibits positive inotropic effects, antiarrhythmic as well as vasoconstrictor actions. It is reported that digoxin and other cardiac glycosides could play important role in the prevention and/or treatment of proliferative diseases like cancer. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 20830-75-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C41H64O14
MDL Number MFCD00003674
Synonym Digoxin; 12 beta-Hydroxydigitoxin
Type Certified Reference Material
Quantity 300 mg
Formula Weight 780.94
Grade Certified Reference Material