accessibility menu, dialog, popup

UserName

BrOOHN

CAS RN 20780-72-7

IUPAC Name: 4-bromo-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 4-bromo-1H-indole-2,3-dione
Molecular Weight (g/mol): 226.03
Molecular Formula: C8H4BrNO2
InChi Key: ITRAKBJPMLKWIW-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1C(=O)C(=O)N2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results