missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoisatin 98.0+%, TCI America™

Catalog No. B24235G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24235G 5 g
B242325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24235G Supplier TCI America Supplier No. B24235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20780-72-7
Molecular Formula C8H4BrNO2
Molecular Weight (g/mol) 226.029
InChI Key ITRAKBJPMLKWIW-UHFFFAOYSA-N
Synonym 4-Bromoindole-2,3-dione, 4-Bromo-2,3-indolinedione
PubChem CID 4500012
IUPAC Name 4-bromo-1H-indole-2,3-dione
SMILES C1=CC2=C(C(=C1)Br)C(=O)C(=O)N2

Specifications

Melting Point 271°C
Quantity 5 g
Formula Weight 226.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromoisatin