CAS RN 207226-31-1
CAS RN 207226-31-1
IUPAC Name:
1,3-dibromo-5-(trifluoromethoxy)benzene
Synonyms:
1,3-dibromo-5-(trifluoromethoxy)benzene
Molecular Weight (g/mol):
319.9
Molecular Formula:
C7H3Br2F3O
InChi Key:
UKHOUWWEEAOCTI-UHFFFAOYSA-N
SMILES:
FC(F)(F)OC1=CC(Br)=CC(Br)=C1