missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-5-(trifluoromethoxy)benzene 98.0+%, TCI America™

Catalog No. D33225G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D33225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D33225G Supplier TCI America Supplier No. D33225G
Only null left

Specifications

CAS 207226-31-1
Color Yellow
Boiling Point 85°C
Molecular Formula C7H3Br2F3O
Quantity 5 g
MDL Number MFCD00075280
Synonym 1,3-dibromo-5-trifluoromethoxy benzene, 1,3-dibromo-5-trifluoromethoxy-benzene, 3,5-dibromotrifluoromethoxybenzene, 3,5-dibromo trifluoromethoxy benzene, 3,5-dibromo-1-trifluoromethoxy benzene, 3,5-dibromo-alpha,alpha,alpha-trifluoroanisole, acmc-1cfab, ksc496c1r, 3,5-dibromo-triffluoromethoxybenzene, 3,5-dibromo-, ,-trifluoroanisole
InChI Key UKHOUWWEEAOCTI-UHFFFAOYSA-N
SMILES FC(F)(F)OC1=CC(Br)=CC(Br)=C1
IUPAC Name 1,3-dibromo-5-(trifluoromethoxy)benzene
Molecular Weight (g/mol) 319.90
PubChem CID 2761179
Formula Weight 319.90
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,3-Dibromo-5-(trifluoromethoxy)benzene
Show More Show Less