accessibility menu, dialog, popup

UserName

CAS RN 20426-12-4

IUPAC Name: (2E)-3-(4-hydroxyphenyl)-1-phenylprop-2-en-1-one
Synonyms: 4-hydroxychalcone
Molecular Weight (g/mol): 224.26
Molecular Formula: C15H12O2
InChi Key: PWWCDTYUYPOAIU-DHZHZOJOSA-N
SMILES: OC1=CC=C(\C=C\C(=O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)