accessibility menu, dialog, popup

UserName

CAS RN 2040-05-3

IUPAC Name: 1-(2,6-dichlorophenyl)ethan-1-one
Synonyms: 1-(2,6-dichlorophenyl)ethanone
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: HYBDSXBLGCQKRE-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(Cl)C=CC=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (1)
  • (5)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)