missing translation for 'onlineSavingsMsg'
Learn More

2',6'-Dichloroacetophenone, 98%

Catalog No. AAL0634806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0634806 5 g
AAL0634803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0634806 Supplier Thermo Scientific Chemicals Supplier No. L0634806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2040-05-3 | C8H6Cl2O | 189.04 g/mol

2',6'-Dichloroacetophenone is is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′,6′-Dichloroacetophenone is is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2040-05-3
MDL Number MFCD00016339

Specifications

Melting Point 41°C to 44°C
Boiling Point 176°C to 180°C (83 mmHg)
Molecular Formula C8H6Cl2O
Quantity 5 g
Beilstein 2440104
Synonym 2',6'-dichloroacetophenone, 1-2,6-dichlorophenyl ethanone, 2,6-dichloroacetophenone, 1-2,6-dichlorophenyl ethan-1-one, ethanone, 1-2,6-dichlorophenyl, 2,6-dichloro acetophenone, 1-acetyl-2,6-dichlorobenzene, zlchem 1084, pubchem12362, acmc-1cobf
Solubility Information Insoluble in water.
InChI Key HYBDSXBLGCQKRE-UHFFFAOYSA-N
SMILES CC(=O)C1=C(C=CC=C1Cl)Cl
IUPAC Name 1-(2,6-dichlorophenyl)ethanone
Molecular Weight (g/mol) 189.04
PubChem CID 74877
Formula Weight 189.04
Percent Purity 98%
Chemical Name or Material 2',6'-Dichloroacetophenone
Show More Show Less
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 218-035-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only