accessibility menu, dialog, popup

UserName

CAS RN 20279-21-4

CAS RN 20279-21-4

IUPAC Name: 1,2,3,10b-tetrahydrofluoranthene
Synonyms: 1,2,3,10b-tetrahydrofluoranthene
Molecular Weight (g/mol): 206.29
Molecular Formula: C16H14
InChi Key: VMBZUGDYWLFLEU-UHFFFAOYNA-N
SMILES: C1CC2C3=CC=CC=C3C3=CC=CC(C1)=C23
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results