accessibility menu, dialog, popup

UserName

NOOBrOFFF

CAS RN 200958-40-3

IUPAC Name: 2-bromo-4-nitro-1-(trifluoromethoxy)benzene
Synonyms: 2-bromo-4-nitro-1-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 286
Molecular Formula: C7H3BrF3NO3
InChi Key: LVTAFGFYMLODQP-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Br)=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results