accessibility menu, dialog, popup

UserName

CAS RN 19932-84-4

IUPAC Name: 6-chloro-2,3-dihydro-1,3-benzoxazol-2-one
Synonyms: 6-chloro-2-benzoxazolinone
Molecular Weight (g/mol): 169.56
Molecular Formula: C7H4ClNO2
InChi Key: MATCZHXABVLZIE-UHFFFAOYSA-N
SMILES: ClC1=CC=C2NC(=O)OC2=C1
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)