missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-2-benzoxazolinone 98.0+%, TCI America™

Catalog No. C23785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23785G 5 g
C237825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23785G Supplier TCI America Supplier No. C23785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19932-84-4
Molecular Formula C7H4ClNO2
Molecular Weight (g/mol) 169.56
MDL Number MFCD00463914
InChI Key MATCZHXABVLZIE-UHFFFAOYSA-N
Synonym 6-chloro-2-benzoxazolinone, 6-chlorobenzoxazolone, 2 3h-benzoxazolone, 6-chloro, 6-chloro-1,3-benzoxazol-2 3h-one, 6-chlorobenzo d oxazol-2 3h-one, 6-chlorobenzoxazolinone, 6-chloro-2-benzoxazolone, 6-chloro-1,3-benzoxazol-2-ol, 2-benzoxazolinone, 6-chloro, 6-chloro-2,3-dihydrobenzoxazol-2-one
PubChem CID 29858
IUPAC Name 6-chloro-2,3-dihydro-1,3-benzoxazol-2-one
SMILES ClC1=CC=C2NC(=O)OC2=C1

Specifications

Melting Point 195°C
Quantity 5 g
Formula Weight 169.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-2-benzoxazolinone
RTECSNumber DM5252000
TSCA No