missing translation for 'onlineSavingsMsg'
Learn More

6-Chloro-1,3-benzoxazol-2(3H)-one, 98+%, Thermo Scientific™

Catalog No. AC297260050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC297260050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC297260050 Supplier Thermo Scientific Chemicals Supplier No. 297260050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19932-84-4
Molecular Formula C7H4ClNO2
Molecular Weight (g/mol) 169.56
MDL Number MFCD00463914
InChI Key MATCZHXABVLZIE-UHFFFAOYSA-N
Synonym 6-chloro-2-benzoxazolinone, 6-chlorobenzoxazolone, 2 3h-benzoxazolone, 6-chloro, 6-chloro-1,3-benzoxazol-2 3h-one, 6-chlorobenzo d oxazol-2 3h-one, 6-chlorobenzoxazolinone, 6-chloro-2-benzoxazolone, 6-chloro-1,3-benzoxazol-2-ol, 2-benzoxazolinone, 6-chloro, 6-chloro-2,3-dihydrobenzoxazol-2-one
PubChem CID 29858
IUPAC Name 6-chloro-3H-1,3-benzoxazol-2-one
SMILES ClC1=CC=C2NC(=O)OC2=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 192.0°C to 193.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: ethanol,methanol,acetic acid,chcl3
Formula Weight 169.57
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-1,3-benzoxazol-2(3H)-one
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protec

RUO – Research Use Only