CAS RN 1986-47-6
IUPAC Name:
hydrogen (1S,2R)-2-phenylcyclopropan-1-amine chloride
Synonyms:
hydrogen (1S,2R)-2-phenylcyclopropan-1-amine chloride
Molecular Weight (g/mol):
169.65
Molecular Formula:
C9H12ClN
InChi Key:
ZPEFMSTTZXJOTM-RJUBDTSPSA-N
SMILES:
[H+].[Cl-].N[C@H]1C[C@@H]1C1=CC=CC=C1
1
–
2
of
2
results
1
Promotions Available
trans-2-Phenylcyclopropylamine hydrochloride, 97%
trans-2-Phenylcyclopropylamine hydrochloride, 97%
CAS: 1986-47-6 Molecular Formula: C9H11N·HCl Molecular Weight (g/mol): 169.66 MDL Number: MFCD00063602 InChI Key: ZPEFMSTTZXJOTM-VTLYIQCISA-N Synonym: tranylcypromine hydrochloride,trans-2-phenylcyclopropanamine hydrochloride,trans-2-phenylcyclopropylamine hydrochloride,1r,2s-2-phenyl-cyclopropylamine hydrochloride,1r,2s-2-phenylcyclopropan-1-amine hydrochloride,tranylcypromine hcl,1r,2s-2-phenylcyclopropanamine hydrochloride,trans-2-phenyl-cyclopropylamine hydrochloride,dsstox_cid_27761,dsstox_rid_82538 PubChem CID: 12345947 IUPAC Name: (1R,2R)-2-phenylcyclopropan-1-amine;hydrochloride SMILES: C1C(C1N)C2=CC=CC=C2.Cl
2
R&D Systems™ Tranylcypromine hydrochloride
CAS: 1986-47-6 Molecular Formula: C9H12ClN