accessibility menu, dialog, popup

UserName

CAS RN 1978-39-8

H3COFH2N

CAS RN 1978-39-8

IUPAC Name: 5-fluoro-2-methoxyaniline
Synonyms: 5-fluoro-2-methoxyaniline
Molecular Weight (g/mol): 141.15
Molecular Formula: C7H8FNO
InChi Key: VYZUBHRSGQAROM-UHFFFAOYSA-N
SMILES: COC1=CC=C(F)C=C1N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results