missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxyaniline 98.0+%, TCI America™

Catalog No. F06145G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F06145G Supplier TCI America Supplier No. F06145G
Only null left

Chemical Identifiers

CAS 1978-39-8
Molecular Formula C7H8FNO
Molecular Weight (g/mol) 141.15
MDL Number MFCD00077537
InChI Key VYZUBHRSGQAROM-UHFFFAOYSA-N
Synonym 2-amino-4-fluoroanisole, 5-fluoro-2-methoxyphenylamine, 5-fluoro-o-anisidine, 5-fluoro-2-methoxybenzenamine, 3-fluoro-6-methoxyaniline, benzenamine, 5-fluoro-2-methoxy, 2-amino-4-fluoro-1-methoxybenzene, 4-fluoro-2-aminoanisole, 2-methoxy-5-fluoroaniline, 2-methoxyl-5-fluoroaniline
PubChem CID 223105
IUPAC Name 5-fluoro-2-methoxyaniline
SMILES COC1=CC=C(F)C=C1N

Specifications

Melting Point 3°C
Color Yellow
Boiling Point 117°C
Quantity 5 g
UN Number 2810
Formula Weight 141.15
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Fluoro-2-methoxyaniline