accessibility menu, dialog, popup

UserName

(R)OHOHNOCH3H3CCH3

CAS RN 19764-30-8

IUPAC Name: (2R)-2-acetamido-4-methylpentanoic acid
Synonyms: D-leucine, N-acetyl-
Molecular Weight (g/mol): 173.21
Molecular Formula: C8H15NO3
InChi Key: WXNXCEHXYPACJF-JLDDOWRYNA-N
SMILES: CC(C)C[C@@H](NC(C)=O)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)