missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-leucine 99.0+%, TCI America™

Catalog No. A07131G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A07131G Supplier TCI America Supplier No. A07131G

Chemical Identifiers

CAS 19764-30-8
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00066069
InChI Key WXNXCEHXYPACJF-JLDDOWRYNA-N
Synonym n-acetyl-d-leucine, ac-d-leu-oh, d-leucine, n-acetyl, n-acethy-d-leucine, n-acetyl-r-leucine, unii-91wu82ga22, 2,n-acetyl-d-leucine, r-2-acetamido-4-methylpentanoic acid, 2r-2-acetamido-4-methylpentanoic acid, acetylleucine, d
PubChem CID 1241420
IUPAC Name (2R)-2-acetamido-4-methylpentanoic acid
SMILES CC(C)C[C@@H](NC(C)=O)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 173.21
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Acetyl-D-leucine