missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-leucine, 99%

Catalog No. AAH2704614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2704614 25 g
AAH2704606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2704614 Supplier Thermo Scientific Chemicals Supplier No. H2704614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 19764-30-8 | C8H15NO3 | 173.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19764-30-8
Molecular Formula C8H15NO3
Molecular Weight (g/mol) 173.21
MDL Number MFCD00066069
InChI Key WXNXCEHXYPACJF-JLDDOWRYNA-N
Synonym n-acetyl-d-leucine, ac-d-leu-oh, d-leucine, n-acetyl, n-acethy-d-leucine, n-acetyl-r-leucine, unii-91wu82ga22, 2,n-acetyl-d-leucine, r-2-acetamido-4-methylpentanoic acid, 2r-2-acetamido-4-methylpentanoic acid, acetylleucine, d
PubChem CID 1241420
IUPAC Name (2R)-2-acetamido-4-methylpentanoic acid
SMILES CC(C)C[C@@H](NC(C)=O)C(O)=O

Specifications

Quantity 25 g
Formula Weight 173.21
Percent Purity 99%
Chemical Name or Material N-Acetyl-D-leucine
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only