accessibility menu, dialog, popup

UserName

CAS RN 196604-20-3

IUPAC Name: 1,3-dimethyl-5-(2-methylphenoxy)benzene
Synonyms: 1,3-dimethyl-5-(2-methylphenoxy)benzene
Molecular Weight (g/mol): 212.29
Molecular Formula: C15H16O
InChi Key: YCAZRHVXMXMPEY-UHFFFAOYSA-N
SMILES: CC1=CC(OC2=CC=CC=C2C)=CC(C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results