accessibility menu, dialog, popup

UserName

OOOHN

CAS RN 19621-92-2

IUPAC Name: 6-oxo-1,6-dihydropyridine-2-carboxylate
Synonyms: 6-oxo-1H-pyridine-2-carboxylate
Molecular Weight (g/mol): 138.1
Molecular Formula: C6H4NO3
InChi Key: VRCWSYYXUCKEED-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC(=O)N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)