missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy-2-pyridinecarboxylic Acid 97.0+%, TCI America™

Catalog No. H08765G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08765G 5 g
H087625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08765G Supplier TCI America Supplier No. H08765G

Chemical Identifiers

CAS 19621-92-2
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00192220
InChI Key VRCWSYYXUCKEED-UHFFFAOYSA-N
Synonym 6-hydroxypicolinic acid, 6-hydroxy-2-pyridinecarboxylic acid, 6-hydroxypyridine-2-carboxylic acid, 6-oxo-1,6-dihydropyridine-2-carboxylic acid, 6-carboxy-2-hydroxy-pyridine, 6-hydroxy-2-pyridine carboxylic acid, 6-hydroxy-2-pyridinecarboxylicacid, 2-pyridinecarboxylic acid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylicacid, 1,6-dihydro-6-oxo, 2-pyridinecarboxylic acid,1,6-dihydro-6-oxo
PubChem CID 242721
IUPAC Name 6-oxo-1H-pyridine-2-carboxylic acid
SMILES C1=CC(=O)NC(=C1)C(=O)O

Specifications

Quantity 5 g
Formula Weight 139.11
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-2-pyridinecarboxylic Acid