accessibility menu, dialog, popup

UserName

CAS RN 19503-26-5

NNNHNNH2HCl

CAS RN 19503-26-5

IUPAC Name: 1H-1,2,4-triazole-1-carboximidamide hydrochloride
Synonyms: 1,2,4-triazole-1-carboximidamide hydrochloride
Molecular Weight (g/mol): 147.57
Molecular Formula: C3H6ClN5
InChi Key: JDDXNENZFOOLTP-UHFFFAOYSA-N
SMILES: Cl.NC(=N)N1C=NC=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)