accessibility menu, dialog, popup

UserName

CAS RN 19500-02-8

IUPAC Name: 3-methoxy-2-methylaniline
Synonyms: 3-methoxy-2-methylaniline
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: OPXLVWLFDKRYRB-UHFFFAOYSA-N
SMILES: COC1=CC=CC(N)=C1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results